missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLC27A4 Recombinant Rabbit Monoclonal Antibody (JE58-29)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA544919
This item is not returnable.
View return policy
Description
SLC27A4 Recombinant Monoclonal Antibody for Western Blot, IHC (P), Flow
This gene encodes a member of a family of fatty acid transport proteins, which are involved in translocation of long-chain fatty acids cross the plasma membrane. This protein is expressed at high levels on the apical side of mature enterocytes in the small intestine, and appears to be the principal fatty acid transporter in enterocytes. Clinical studies suggest this gene as a candidate gene for the insulin resistance syndrome. Mutations in this gene have been associated with ichthyosis prematurity syndrome.
Specifications
| SLC27A4 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q6P1M0, Q91VE0 | |
| SLC27A4 | |
| Recombinant protein within human SLC27A4 aa 393-643. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| JE58-29 | |
| Unconjugated | |
| SLC27A4 | |
| ACSVL4; BB144259; Fatp4; FATP-4; fatty acid transport protein 4; IPS; Long-chain fatty acid transport protein 4; SLC27A4; solute carrier family 27 (fatty acid transporter), member 4; solute carrier family 27 member 4 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 10999, 26569, 311839 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction