missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC27A5 Mouse anti-Human, Clone:4B11C10, Abnova™
Mouse monoclonal antibody raised against partial recombinant human SLC27A5.
Brand: Abnova MAB17883.100ug
This item is not returnable.
View return policy
Description
The protein encoded by this gene is an isozyme of very long-chain acyl-CoA synthetase (VLCS). It is capable of activating very long-chain fatty-acids containing 24- and 26-carbons. It is expressed in liver and associated with endoplasmic reticulum but not with peroxisomes. Its primary role is in fatty acid elongation or complex lipid synthesis rather than in degradation. This gene has a mouse ortholog. [provided by RefSeq]Specifications
| SLC27A5 | |
| Monoclonal | |
| Unconjugated | |
| ELISA (1:10000) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) Western Blot (1:100-1:500) The optimal working dilution should be determined by the end user. |
|
| SLC27A5 | |
| Mouse | |
| 100 μg | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 4B11C10 | |
| solute carrier family 27 (fatty acid transporter), member 5 | |
| In PBS (0.05% sodium azide) | |
| ACSB, ACSVL6, FACVL3, FATP5, FLJ22987, VLACSR, VLCS-H2, VLCSH2 | |
| Recombinant protein corresponding to amino acids 508-570 of human SLC27A5. | |
| 10998 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction