missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SLC27A6 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17161414 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17161414 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17161414 Supplier Invitrogen™ Supplier No. PA5113556

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SLC27A6 Polyclonal Antibody for Western Blot, ELISA

The solute carrier family 27 (fatty acid transporter), member 6 (SLC27A6) is a member of the fatty acid transport protein family (FATP), which are involved in the uptake of long-chain fatty acids. SLC27A6 is principally expressed in the heart on the sarcolemma and colocalizes with CD36, another molecule involved in the uptake of long chain fatty acids.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC27A6
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.57 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene SLC27A6
Gene Accession No. Q9Y2P4
Gene Alias 4732438L20Rik; ACSVL2; acyl-CoA synthetase very long chain family, member 2; FACVL2; FATP1; FATP6; FATP-6; Fatty-acid-coenzyme A ligase, very long-chain 2; long-chain fatty acid transport protein 6; SLC27A6; solute carrier family 27 (fatty acid transporter), member 6; solute carrier family 27 member 6; Very long-chain acyl-CoA synthetase homolog 1; VLCSH1; VLCS-H1
Gene Symbols SLC27A6
Host Species Rabbit
Immunogen Recombinant Human Long-chain fatty acid transport protein 6 protein (370-619AA).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 28965
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.