missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC29A3 Antibody, DyLight 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-81829DL594
This item is not returnable.
View return policy
Description
SLC29A3 Polyclonal antibody specifically detects SLC29A3 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| SLC29A3 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| DyLight 594 | |
| equilibrative nucleoside transporter 3, ENT3, HCLAP, HJCD, PHID, solute carrier family 29 (nucleoside transporters), member 3 | |
| SLC29A3 antibody was raised against a 19 amino acid peptide near the amino terminus of human SLC29A3. The immunogen is located within the first 50 amino acids of SLC29A3. Amino Acid Squence: LEKLLDRPPPGLQRPEDRF | |
| 0.1 mL | |
| Cancer | |
| 55315 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction