missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC30A1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-95127-0.1ml
This item is not returnable.
View return policy
Description
SLC30A1 Polyclonal antibody specifically detects SLC30A1 in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| SLC30A1 | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunohistochemistry-Paraffin | |
| solute carrier family 30 (zinc transporter), member 1, Solute carrier family 30 member 1, zinc transporter 1, ZnT-1, ZNT1znT-1, ZRC1 | |
| A synthetic peptide corresponding to a sequence within amino acids 251-350 of human SLC30A1 (NP_067017.2). HVLGDALGSVIVVVNALVFYFSWKGCSEGDFCVNPCFPDPCKAFVEIINSTHASVYEAGPCWVLYLDPTLCVVMVCILLYTTYPLLKESALILLQTVPKQ | |
| 0.1 mL | |
| Primary | |
| Human, Rat | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 7779 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction