missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLC30A10 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA549188
This item is not returnable.
View return policy
Description
SLC30A10 Polyclonal Antibody for Western Blot
May be involved in zinc transport out of the cell, being a zinc-efflux transporter.
Specifications
| SLC30A10 | |
| Polyclonal | |
| Unconjugated | |
| SLC30A10 | |
| E130106K10Rik; Gm212; HMDPC; HMNDYT1; Manganese transporter SLC30A10; SLC30A10; Solute carrier family 30 member 10; solute carrier family 30, member 10; zinc transporter 10; zinc transporter 8; Znt10; znT-10; ZNT8; ZRC1 | |
| Rabbit | |
| Antigen Affinity Chromatography, Protein A | |
| RUO | |
| 55532 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q6XR72 | |
| SLC30A10 | |
| KLH conjugated synthetic peptide between 331-359 amino acids of human SLC30A10. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction