missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SLC30A3 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15979614
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15979614 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15979614 Supplier Invitrogen™ Supplier No. PA566938

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: GLETTRLVSPR DRGGAGGSLR LKSLFTEPSE PLPEESKPVE MPFHHCHRDP LPPPGLTPER LHARRQL Highest antigen sequence identity to the following orthologs - mouse 75%, rat 71%.

Slc30a3 encodes a protein that is involved in accumulation of zinc in synaptic vesicles.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC30A3
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Slc30a3
Gene Accession No. Q99726
Gene Alias SLC30A3; solute carrier family 30 (zinc transporter), member 3; solute carrier family 30 member 3; solute carrier family 30 protein; zinc transporter 3; zinc transporter ZnT-3; ZNT3; znT-3
Gene Symbols Slc30a3
Host Species Rabbit
Immunogen Recombinant Human SLC30A3. Recombinant protein control fragment (Product #RP-107591).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7781
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.