missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC30A5, Mouse anti-Human, Clone: 2E10, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00064924-M03.50ug
This item is not returnable.
View return policy
Description
Sequence: MEEKYGGDVLAGPGGGGGLGPVDVPSARLTKYIVLLCFTKFLKAVGLFESYDLLKAVHIVQFIFILKLGTAFFMVLFQKPFSSGKTITKHQIIGSLKIPGRKEFKDKKLNDPRKLVGNSpecifications
| SLC30A5 | |
| Monoclonal | |
| Unconjugated | |
| solute carrier family 30 (zinc transporter), member 5 | |
| FLJ12496/FLJ12756/MGC5499/ZNT5/ZNTL1/ZTL1/ZnT-5 | |
| Mouse | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Western Blot | |
| 2E10 | |
| In 1x PBS, pH 7.4 | |
| BC000808 | |
| SLC30A5 | |
| SLC30A5 (AAH00808.1, 1 a.a. ∼ 118 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 64924 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction