missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLC30A6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA563432
This item is not returnable.
View return policy
Description
Immunogen sequence: NVNPVILLNT QTRPYGFGLN HGHTPYSSML NQGLGVPGIG ATQGLRTGFT NIPSRYGTNN RIGQPRP Highest antigen sequence identity to the following orthologs: Mouse - 73%, Rat - 76%.
Zinc functions as a cofactor for numerous enzymes, nuclear factors, and hormones and as an intra- and intercellular signal ion. Members of the zinc transporter (ZNT)/SLC30 subfamily of the cation diffusion facilitator family, such as SLC30A6, permit cellular efflux of zinc (Seve et al., 2004).
Specifications
| SLC30A6 | |
| Polyclonal | |
| Unconjugated | |
| SLC30A6 | |
| 9530029F08Rik; MST103; MSTP103; SLC30A6; solute carrier family 30 (zinc transporter), member 4; solute carrier family 30 (zinc transporter), member 6; solute carrier family 30 member 6; Zinc transporter 6; zinc transporter-like 3; Znt6; znT-6 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 55676 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q6NXT4 | |
| SLC30A6 | |
| Recombinant protein corresponding to Human SLC30A6. Recombinant protein control fragment (Product #RP-95655). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction