missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLC30A9 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA552043
This item is not returnable.
View return policy
Description
Immunogen sequence: HRCSWSSLCR LRLRCRAAAC NPSDRQEWQN LVTFGSFSNV VPCSHPYIGT LSQVKLYSTN VQKEGQGSQT LRVEKVPSFE TAEGIGAELK APLKQEPLQV RVKAVLKKRE YGSKYTQNNF ITGV Highest antigen sequence identity to the following orthologs: Mouse - 66%, Rat - 66%.
The gene corresponding to embryonic lung protein [also known as Solute carrier family 30 (Zinc transporter), member 9, SLC30A9] is likely to be an evolutionarily conserved, housekeeping gene that plays a role intimately linked with cellular replication, DNA synthesis and/or transcriptional regulation.
Specifications
| SLC30A9 | |
| Polyclonal | |
| Unconjugated | |
| SLC30A9 | |
| 2310024J23Rik; AL024256; C4orf1; expressed in human embryonic lung; GAC63; GRIP1-associated coactivator 1; GRIP1-associated coactivator 63; GRIP1-dependent nuclear receptor coactivator; HUEL; Human embryonic lung protein; Slc30a9; solute carrier family 30 (zinc transporter), member 9; solute carrier family 30 member 9; Zinc transporter 9; ZNT9; znT-9 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10463 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q6PML9 | |
| SLC30A9 | |
| Recombinant protein corresponding to Human SLC30A9. Recombinant protein control fragment (Product #RP-102351). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction