missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC34A1 Antibody (10B1.3E9), PE/Atto594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-42216PEATT594
This item is not returnable.
View return policy
Description
SLC34A1 Monoclonal specifically detects SLC34A1 in Human, Mouse samples. It is validated for Flow Cytometry.
Specifications
| SLC34A1 | |
| Monoclonal | |
| PE/ATTO 594 | |
| PBS with 0.05% Sodium Azide | |
| SLC34A1 | |
| Two synthetic peptides made to an internal portions of the human SLC34A1 protein (between amino acids 25-150 and 100-200) [UniProt Q06495] | |
| 0.1 mL | |
| 6569.0 | |
| Store at 4°C in the dark. Do not freeze. |
| Flow Cytometry | |
| 10B1.3E9 | |
| Flow Cytometry | |
| FRTS2, Na(+)/Pi cotransporter 2A, Na(+)-dependent phosphate cotransporter 2A, NaPi-2a, NaPi-3, NPT2NPHLOP1, NPTIIa, renal sodium-dependent phosphate transporter, SLC11, SLC17A2Na+-phosphate cotransporter type II, sodium/phosphate co-transporter, Sodium/phosphate cotransporter 2A, sodium-dependent phosphate transport protein 2A, Sodium-phosphate transport protein 2A, solute carrier family 17 (sodium phosphate), member 2, solute carrier family 34 (sodium phosphate), member 1, Solute carrier family 34 member 1 | |
| Mouse | |
| Protein G purified | |
| Primary | |
| Human, Mouse | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction