missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ SLC34A2 Antibody (Lifastuzumab), Novus Biologicals™
Human Monoclonal Antibody
€ 213.00 - € 920.00
Specifications
| Antigen | SLC34A2 |
|---|---|
| Clone | Lifastuzumab |
| Applications | Flow Cytometry, ELISA, Functional Assay |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30048014
|
Novus Biologicals™
NBP3-28345-50ug |
50 μg |
€ 213.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30048693
|
Novus Biologicals™
NBP3-28345-1mg |
1 mg |
€ 920.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30045054
|
Novus Biologicals™
NBP3-28345-100ug |
100 μg |
€ 349.00 € 329.70 / 100µg Save € 19.30 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SLC34A2 Monoclonal antibody specifically detects SLC34A2 in Human samples. It is validated for Flow Cytometry,ELISA,Functional AssaySpecifications
| SLC34A2 | |
| Flow Cytometry, ELISA, Functional Assay | |
| Unconjugated | |
| Human | |
| Endocrinology, Signal Transduction | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| NAPI-3B, NPTIIb, sodium/phosphate cotransporter 2B, solute carrier family 34 (sodium phosphate), member 2, type II sodium-dependent phosphate transporter 3b | |
| NaPi2b/ SLC34A2 | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. |
| Lifastuzumab | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| O95436 | |
| 10568 | |
| IgG1 | |
| Protein A purified | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title