missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC35A1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16342-1-AP-20UL
This item is not returnable.
View return policy
Specifications
| SLC35A1 | |
| Polyclonal | |
| Unconjugated | |
| SLC35A1 | |
| CMP SA Tr, CMP Sia Tr, CMP sialic acid transporter, CMPST, CST, hCST, inactive, SLC35A1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10559, 24060, 313139 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation | |
| 0.2 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P78382, Q61420 | |
| Slc35a1 | |
| SLC35A1 Fusion Protein Ag9524 | |
| 20 μL | |
| Primary | |
| Mouse, Rat, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction