missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SLC35C1 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA564146

Product Code. 15761912

  • € 582.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: WVRGWEMKKT PEEPSPKDSE KSAMGV Highest antigen sequence identity to the following orthologs: Mouse - 73%, Rat - 73%.

SLC35C1 is involved in GDP-fucose import from the cytoplasm into the Golgi lumen.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

SLC35C1
Polyclonal
Unconjugated
SLC35C1
CDG2C; E430007K15Rik; fuct1; GDP-fucose transporter; GDP-fucose transporter 1; GDP-Fuc-Tr; golgi GDP-fucose translocator; multispan transmembrane protein fuct1; putative golgi GDP-fucose translocator; SLC35C1; solute carrier family 35 (GDP-fucose transporter), member C1; solute carrier family 35 member C1; solute carrier family 35, member C1
Rabbit
Antigen affinity chromatography
RUO
55343
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Immunocytochemistry
0.05 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q96A29
SLC35C1
Recombinant protein corresponding to Human SLC35C1. Recombinant protein control fragment (Product #RP-104180).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.