missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Immunogen sequence: RYQFSFLTLV QCLTSSTAAL SLELLRRLGL IAVPPFGLSL ARSFAGVAVL For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 79%.
May play a role in hemostasis as a regulator of the biosynthesis of platelet-dense granules.
Specifications
Specifications
| Antigen | SLC35D3 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 2% sucrose and 0.09% sodium azide |
| Gene | SLC35D3 |
| Gene Accession No. | Q5M8T2 |
| Gene Alias | 6230421J19Rik; frc, fringe-like 1; Frcl1; Fringe connection-like protein 1; SLC35D3; Solute carrier family 35 member D3; solute carrier family 35, member D3 |
| Gene Symbols | SLC35D3 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
