missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC35E1 Antibody, DyLight 680, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97796FR
This item is not returnable.
View return policy
Description
SLC35E1 Polyclonal antibody specifically detects SLC35E1 in Human samples. It is validated for Western Blot
Specifications
| SLC35E1 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 79939 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| DyLight 680 | |
| FLJ14251, FLJ36689, solute carrier family 35 member E1, solute carrier family 35, member E1 | |
| Produced in rabbits immunized with E. coli-derived Human SLC35E1 fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction