missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLC35E2B Polyclonal Antibody

Description
This target displays homology in the following species: Dog: 79%; Human: 100%.
Putative transporter.
Specifications
Specifications
| Antigen | SLC35E2B |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 2% sucrose and 0.09% sodium azide |
| Gene | SLC35E2B |
| Gene Accession No. | P0CK96 |
| Gene Alias | KIAA0447; SLC35E2; SLC35E2B; Solute carrier family 35 member E2B; solute carrier family 35, member E2B |
| Gene Symbols | SLC35E2B |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?