Learn More
Invitrogen™ SLC35F5 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA598692
Product Code. 16571512
  • € 464.00 / 100µg
Description
SLC35F5 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA
SLC35F5 is a putative solute transporter.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SLC35F5
Polyclonal
Unconjugated
SLC35F5
1300003P13Rik; AI646727; HCV NS5A-transactivated protein 3; hepatitis C virus NS5A-transactivated protein 3; NS5ATP3; SLC35F5; Solute carrier family 35 member F5; solute carrier family 35, member F5; UNQ2545/PRO6097
Rabbit
Protein G
RUO
80255
-20°C or -80°C if preferred
Liquid
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
3 mg/mL
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Q8WV83
SLC35F5
Recombinant Human Solute carrier family 35 member F5 protein (122-242AA).