Learn More
SLC35G2 Antibody, PE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97661PE
This item is not returnable.
View return policy
Description
SLC35G2 Polyclonal antibody specifically detects SLC35G2 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence
Specifications
| SLC35G2 | |
| Polyclonal | |
| Immunocytochemistry/ Immunofluorescence | |
| Produced in rabbits immunized with E. coli-derived Human SLC35G2 fragment. | |
| Primary | |
| Human |
| Immunocytochemistry | |
| PE | |
| Rabbit | |
| 0.1 mL | |
| 80723 | |
| Store at 4C in the dark. |