missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC36A2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 391.65 - € 590.10
Specifications
| Antigen | SLC36A2 |
|---|---|
| Dilution | Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18639218
|
Novus Biologicals
NBP2-49360 |
0.1 mL |
€ 624.00 € 590.10 / 0.10mL Save € 33.90 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18663759
|
Novus Biologicals
NBP2-49360-25ul |
25 μL |
€ 415.00 € 391.65 / 25µL Save € 23.35 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SLC36A2 Polyclonal antibody specifically detects SLC36A2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| SLC36A2 | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Neuroscience | |
| PBS (pH 7.2), 40% Glycerol | |
| 153201 | |
| IgG | |
| Immunogen affinity purified |
| Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| FLJ16051, MGC119658, MGC119660, PAT2proton-coupled amino acid transporter 2, Proton/amino acid transporter 2, solute carrier family 36 (proton/amino acid symporter), member 2, Solute carrier family 36 member 2, TRAMD1tramdorin-1, tramdorin, Tramdorin-1 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: QALDELLKSEDSHPFSNSTTFVR | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title