missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
SLC37A4 transports glucose-6-phosphate from the cytoplasm to the lumen of the endoplasmic reticulum. It forms with glucose-6-phosphatase the complex responsible for glucose production through glycogenolysis and gluconeogenesis. Hence, it plays a central role in homeostatic regulation of blood glucose levels.
Specifications
Specifications
| Antigen | SLC37A4 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.18 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | SLC37A4 |
| Gene Accession No. | O43826 |
| Gene Alias | G6PT, G6PT1, G6PT2, G6PT3, GSD1b, GSD1c, GSD1d, PRO0685, SLC37A4, TRG 19, TRG19 |
| Gene Symbols | SLC37A4 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?