missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC39A10 Antibody, DyLight 550, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76507R
This item is not returnable.
View return policy
Description
SLC39A10 Polyclonal antibody specifically detects SLC39A10 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| SLC39A10 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| DyLight 550 | |
| DKFZp564L2123, DKFZp781L10106, FLJ90515, KIAA1265, LZT-Hs2, MGC126565, MGC138428, solute carrier family 39 (metal ion transporter), member 10, solute carrier family 39 (zinc transporter), member 10, Solute carrier family 39 member 10, zinc transporter ZIP10, ZIP10, ZIP-10, Zrt- and Irt-like protein 10 | |
| Antibody was raised against an 18 amino acid synthetic peptide near the center of human ZIP10. The immunogen is located within amino acids 550 - 600 of ZIP10. Amino Acid Squence: AGTDDSVVSEDRLNETEL | |
| 0.1 mL | |
| Cancer, Cell Cycle and Replication | |
| 57181 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction