missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC39A13 Antibody, FITC, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76509F
This item is not returnable.
View return policy
Description
SLC39A13 Polyclonal antibody specifically detects SLC39A13 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| SLC39A13 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| FITC | |
| FLJ25785, LIV-1 subfamily of ZIP zinc transporter 9, LZT-Hs9, solute carrier family 39 (metal ion transporter), member 13, solute carrier family 39 (zinc transporter), member 13, Solute carrier family 39 member 13, zinc transporter ZIP13, ZIP13, ZIP-13, Zrt- and Irt-like protein 13 | |
| Antibody was raised against a 14 amino acid synthetic peptide near the amino terminus of human ZIP13. The immunogen is located within amino acids 150 - 200 of ZIP13. Amino Acid Squence: DSKEEKTSQAPNKD | |
| 0.1 mL | |
| Signal Transduction | |
| 91252 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction