missing translation for 'onlineSavingsMsg'
Learn More

SLC39A14 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP1-76510-0.025mg

 View more versions of this product

Product Code. 18572001

  • € 189.00 / 25µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

SLC39A14 Polyclonal specifically detects SLC39A14 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

SLC39A14
Polyclonal
Unconjugated
PBS with 0.02% Sodium Azide
Irt-like protein 14, LIV-1 subfamily of ZIP zinc transporter 4, LZT-Hs4, solute carrier family 39 (metal ion transporter), member 14, solute carrier family 39 (zinc transporter), member 14, solute carrier family 39 member 14, zinc transporter ZIP14, ZIP14, ZIP-14, zrt- and Irt-like protein 14
Rabbit
52 kDa
0.025 mg
Primary
At least three isoforms of ZIP14 are known to exist; this antibody will detect both isoforms. ZIP14 antibody is predicted to not cross-react with other ZIP family members.
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
Western Blot
1 mg/mL
Western Blot 1-2 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 2.5 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg/mL., Immunohistochemistry-Paraffin 2.5 μg/mL
NP_001128625
SLC39A14
Antibody was raised against a 16 amino acid synthetic peptide near the center of human ZIP14. The immunogen is located within amino acids 230 - 280 of ZIP14. Amino Acid Squence: KQKNEHHHGHSHYASE
Affinity Purified
RUO
23516
Human
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only