missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC39A14 Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76510H
This item is not returnable.
View return policy
Description
SLC39A14 Polyclonal antibody specifically detects SLC39A14 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| SLC39A14 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| HRP | |
| Irt-like protein 14, LIV-1 subfamily of ZIP zinc transporter 4, LZT-Hs4, solute carrier family 39 (metal ion transporter), member 14, solute carrier family 39 (zinc transporter), member 14, solute carrier family 39 member 14, zinc transporter ZIP14, ZIP14, ZIP-14, zrt- and Irt-like protein 14 | |
| Antibody was raised against a 16 amino acid synthetic peptide near the center of human ZIP14. The immunogen is located within amino acids 230 - 280 of ZIP14. Amino Acid Squence: KQKNEHHHGHSHYASE | |
| 0.1 mL | |
| Cancer, Endocrinology, Signal Transduction | |
| 23516 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction