missing translation for 'onlineSavingsMsg'
Learn More

SLC39A14 Antibody, mFluor Violet 500 SE, Novus Biologicals™

Product Code. 18462708 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18462708 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18462708 Supplier Novus Biologicals Supplier No. NBP176510MFV500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SLC39A14 Polyclonal antibody specifically detects SLC39A14 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC39A14
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate mFluor Violet 500 SE
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias Irt-like protein 14, LIV-1 subfamily of ZIP zinc transporter 4, LZT-Hs4, solute carrier family 39 (metal ion transporter), member 14, solute carrier family 39 (zinc transporter), member 14, solute carrier family 39 member 14, zinc transporter ZIP14, ZIP14, ZIP-14, zrt- and Irt-like protein 14
Host Species Rabbit
Immunogen Antibody was raised against a 16 amino acid synthetic peptide near the center of human ZIP14. The immunogen is located within amino acids 230 - 280 of ZIP14. Amino Acid Squence: KQKNEHHHGHSHYASE
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Endocrinology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 23516
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.