Learn More
Invitrogen™ SLC39A2 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA599743
Product Code. 16693985
  • € 493.00 / 100µL
Description
Antibody detects endogenous levels of total ZIP2.
The zinc transporter ZIP2, also known as SLC39A2, is a member of a family of divalent ion transporters. Zinc is an essential ion for cells and plays significant roles in the growth, development, and differentiation. Similar to knock-outs of ZIP1 and ZIP3, ZIP2-null mice have no phenotypic differences compared to wild-type mice. Only when ZIP1, ZIP2, and ZIP3 genes are all eliminated and these mutant mice are fed a zinc-deficient diet do abnormalities such as reduced embryonic-membrane bound alkaline phosphatase activity and abnormal development occur, indicating that the ZIP1-3 proteins play an important, noncompensatory role when zinc is deficient. More recent studies have shown that ZIP2 and ZIP3 are down regulated in human prostate adenocarcinomatous glands, and may be important in the retention of zinc in the cellular compartment.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SLC39A2
Polyclonal
Unconjugated
SLC39A2
6A1; ETI-1; F730005G13Rik; Gm1789; Gm289; MGC119190; SLC39A2; solute carrier family 39 (zinc transporter), member 2; solute carrier family 39 member 2; Zinc transporter ZIP2; zinc uptake transporter 2; ZIP2; ZIP-2; Zrt- and Irt-like protein 2
Rabbit
Affinity chromatography
RUO
29986
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q9NP94
SLC39A2
A synthesized peptide derived from human SLC39A2(Accession Q9NP94), corresponding to amino acid residues A14-M64.