missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLC39A2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA556486
This item is not returnable.
View return policy
Description
Immunogen sequence: QCCPGAAGGS TVQDEEWGGA HIFELHSHGH LPSPS Highest antigen sequence identity to the following orthologs: Mouse - 71%, Rat - 71%.
The zinc transporter ZIP2, also known as SLC39A2, is a member of a family of divalent ion transporters. Zinc is an essential ion for cells and plays significant roles in the growth, development, and differentiation. Similar to knock-outs of ZIP1 and ZIP3, ZIP2-null mice have no phenotypic differences compared to wild-type mice. Only when ZIP1, ZIP2, and ZIP3 genes are all eliminated and these mutant mice are fed a zinc-deficient diet do abnormalities such as reduced embryonic-membrane bound alkaline phosphatase activity and abnormal development occur, indicating that the ZIP1-3 proteins play an important, noncompensatory role when zinc is deficient. More recent studies have shown that ZIP2 and ZIP3 are down regulated in human prostate adenocarcinomatous glands, and may be important in the retention of zinc in the cellular compartment.
Specifications
| SLC39A2 | |
| Polyclonal | |
| Unconjugated | |
| SLC39A2 | |
| 6A1; ETI-1; F730005G13Rik; Gm1789; Gm289; MGC119190; SLC39A2; solute carrier family 39 (zinc transporter), member 2; solute carrier family 39 member 2; Zinc transporter ZIP2; zinc uptake transporter 2; ZIP2; ZIP-2; Zrt- and Irt-like protein 2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 29986 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9NP94 | |
| SLC39A2 | |
| Recombinant protein corresponding to Human SLC39A2. Recombinant protein control fragment (Product #RP-89135). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction