missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC39A2/ZIP2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76499
This item is not returnable.
View return policy
Description
SLC39A2/ZIP2 Polyclonal specifically detects SLC39A2/ZIP2 in Human, Mouse, Rat, Equine samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| SLC39A2/ZIP2 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| Eti-1, MGC119190, solute carrier family 39 (zinc transporter), member 2, Solute carrier family 39 member 2, zinc transporter ZIP2, ZIP-2, ZIP2hZIP2, Zrt- and Irt-like protein 2,6A1 | |
| Rabbit | |
| 32 kDa | |
| 0.1 mg | |
| Primary | |
| Human, Mouse, Rat, Equine | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| Western Blot 1 ug/ml, ELISA 1:100-1:2000, Immunohistochemistry 2.5 ug/ml, Immunocytochemistry/Immunofluorescence 20 ug/ml, Immunohistochemistry-Paraffin 2.5 ug/mL | |
| NP_055394 | |
| SLC39A2 | |
| Antibody was raised against a 17 amino acid synthetic peptide near the amino terminus of human ZIP2. The immunogen is located within amino acids 70 - 120 of ZIP2. Amino Acid Squence: NRSASERNSSGDADSAH | |
| Affinity Purified | |
| RUO | |
| 29986 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur