missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC39A5 Monoclonal Antibody (OTI2F2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA810144S
This item is not returnable.
View return policy
Description
SLC39A5 Monoclonal antibody specifically detects SLC39A5 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| SLC39A5 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q99607 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 2000 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI2F2 | |
| Unconjugated | |
| Elf4 | |
| 1810013D05Rik; 2010205A06Rik; LZT-Hs7; MGC34778; MYP24; SLC39A5; solute carrier family 39 (metal ion transporter), member 5; solute carrier family 39 (zinc transporter), member 5; solute carrier family 39 member 5; Zinc transporter ZIP5; Zip5; ZIP-5; zrt- and Irt-like protein 5 | |
| Human recombinant protein fragment corresponding to amino acids 83-345 of human ELF4 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction