missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC39A8/ZIP8 Antibody, Biotin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97997B
This item is not returnable.
View return policy
Description
SLC39A8/ZIP8 Polyclonal specifically detects SLC39A8/ZIP8 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| SLC39A8/ZIP8 | |
| Polyclonal | |
| Immunohistochemistry, Immunohistochemistry-Paraffin | |
| BCG induced integral membrane protein BIGM103, BIGM103BCG-induced integral membrane protein in monocyte clone 103 protein, LIV-1 subfamily of ZIP zinc transporter 6, LZT-Hs6, solute carrier family 39 (metal ion transporter), member 8, solute carrier family 39 (zinc transporter), member 8, Solute carrier family 39 member 8, zinc transporter ZIP8, ZIP8, ZIP-8, Zrt- and Irt-like protein 8 | |
| Produced in rabbits immunized with E. coli-derived Human SLC39A8/ZIP8 fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Biotin | |
| PBS | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| 64116 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction