missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SLC41A3 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15614695
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15614695 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15614695 Supplier Invitrogen™ Supplier No. PA550763

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

The antibody detects endogenous levels of total SLC41A3 protein.

SLC41A3 (solute carrier family 41 member 3) is a 507 amino acid multi-pass membrane protein that belongs to the SLC41A transporter family that includes SLC41A1 and SLC41A2. Localizing to the cell membrane, SLC41A3 contains ten transmembrane domains and is believed to function as a plasma-membrane cation transporter. Its other two family members, SLC41A1 and SLC41A2, have been characterized to function as magnesium transporters and are essential for maintaining magnesium homeostasis within the cell.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC41A3
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 3.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Gene SLC41A3
Gene Accession No. Q96GZ6
Gene Alias SLC41A1-L2; SLC41A1-like 2; SLC41A3; Solute carrier family 41 member 3; solute carrier family 41, member 3
Gene Symbols SLC41A3
Host Species Rabbit
Immunogen Synthetic peptide corresponding to a region derived from internal residues of human solute carrier family 41, member 3.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54946
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.