Learn More
Invitrogen™ SLC44A1 Monoclonal Antibody (VIM-15b), FITC

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA528190
Description
Mouse anti human CD92 antibody, clone VIM-15b recognizes the C-terminal variant of human CDw92 (isoform 3), also known as CTL1, a 70 kDa multi-pass membrane protein, expressed by monocytes, neutrophils, certain myeloid and T cell lines, and weakly by endothelial cells, fibroblasts and epithelial cells.
SLC44A1 encodes a choline transporter and may be involved in membrane synthesis and myelin production.
Specifications
| SLC44A1 | |
| Monoclonal | |
| 100 μg/mL | |
| PBS with 1% BSA and 0.09% sodium azide | |
| Q8WWI5 | |
| Slc44a1 | |
| MV4-11 acute monocyte leukemia cells. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Flow Cytometry | |
| VIM-15b | |
| FITC | |
| Slc44a1 | |
| 2210409B22Rik; 4833416H08Rik; AW547365; CD92; CDW92; CDW92 antigen; choline transporter-like protein 1; CHTL1; Ctl1; Slc44a1; solute carrier family 44 (choline transporter), member 1; solute carrier family 44 member 1; solute carrier family 44, member 1; transporter-like protein | |
| Mouse | |
| Protein G | |
| RUO | |
| 23446 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.