missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLC44A1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA566711
This item is not returnable.
View return policy
Description
Immunogen sequence: LFLCLAIDTKY NDGSPGREFY MDKVLMEFVE NSRKAMKEAG KGGVADSREL KPMASGASS Highest antigen sequence identity to the following orthologs - mouse 95%, rat 95%.
SLC44A1 encodes a choline transporter and may be involved in membrane synthesis and myelin production.
Specifications
| SLC44A1 | |
| Polyclonal | |
| Unconjugated | |
| Slc44a1 | |
| 2210409B22Rik; 4833416H08Rik; AW547365; CD92; CDW92; CDW92 antigen; choline transporter-like protein 1; CHTL1; Ctl1; Slc44a1; solute carrier family 44 (choline transporter), member 1; solute carrier family 44 member 1; solute carrier family 44, member 1; transporter-like protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23446 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.4 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8WWI5 | |
| Slc44a1 | |
| Recombinant Human SLC44A1. Recombinant protein control fragment (Product #RP-107364). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction