missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SLC44A3 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15749331 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15749331 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15749331 Supplier Invitrogen™ Supplier No. PA561517

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: GAAGRLLFGY DSFGNMCGKK NSPVEGAPLS GQDMTLKKHV FFMNSCNLEV KGTQLNRMAL CVSNCPEEQL DSLEEVQ Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 84%.

SLC44A3 is a multi-pass membrane protein. It belongs to the CTL (choline transporter-like) family. The function of the RBM34 protein remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC44A3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SLC44A3
Gene Accession No. Q8N4M1
Gene Alias BC010552; choline transporter-like protein 3; Ctl3; RGD1305808; SLC44A3; Solute carrier family 44 member 3; solute carrier family 44, member 3; UNQ558/PRO1115
Gene Symbols SLC44A3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SLC44A3. Recombinant protein control fragment (Product #RP-99788).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 126969
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.