missing translation for 'onlineSavingsMsg'
Learn More

SLC45A3/Prostein Antibody (SLC45A3/4965R), Allophycocyanin, Novus Biologicals™

Product Code. 18662427 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18662427 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18662427 Supplier Novus Biologicals Supplier No. NBP314048APC

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

SLC45A3/Prostein Monoclonal antibody specifically detects SLC45A3/Prostein in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC45A3/Prostein
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone SLC45A3/4965R
Conjugate APC
Formulation PBS
Gene Alias IPCA-2, IPCA-6, IPCA-8, PCANAP6IPCA6, PCANAP8, prostate cancer associated protein 2, prostate cancer associated protein 6, prostate cancer associated protein 8, prostate cancer-associated gene 2, prostate cancer-associated gene 6, prostate cancer-associated gene 8, Prostate cancer-associated protein 6, Prostein, PRSTPCANAP2, solute carrier family 45 member 3, solute carrier family 45, member 3
Host Species Rabbit
Immunogen Synthesized peptides to the N terminus of human SLC45A3/Prostein protein (Uniprot: Q96JT2)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 85414
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.