missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC45A3/Prostein Rabbit anti-Human, Clone: ZR9, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-07403-100ug
This item is not returnable.
View return policy
Description
SLC45A3/Prostein Monoclonal specifically detects SLC45A3/Prostein in Human samples. It is validated for Immunohistochemistry-Paraffin, Protein Array.
Specifications
| SLC45A3/Prostein | |
| Monoclonal | |
| 0.2 mg/mL | |
| Immunohistochemistry-Paraffin 1 to 2 μg/mL, Protein Array | |
| IPCA-2, IPCA-6, IPCA-8, PCANAP6IPCA6, PCANAP8, prostate cancer associated protein 2, prostate cancer associated protein 6, prostate cancer associated protein 8, prostate cancer-associated gene 2, prostate cancer-associated gene 6, prostate cancer-associated gene 8, Prostate cancer-associated protein 6, Prostein, PRSTPCANAP2, solute carrier family 45 member 3, solute carrier family 45, member 3 | |
| Synthesized peptides to the N terminus of human SLC45A3/Prostein protein | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin), Peptide Array | |
| ZR9 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| 85414 | |
| Store at 4°C. | |
| IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction