missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Rat Connexin 31/GJB3 ELISA Kit (Colorimetric)

Product Code. 18218927 Shop All Bio Techne Products
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 18218927

Brand: Novus Biologicals™ NBP279815

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This kit recognizes Rat CX31 in samples.

No Significant cross-reactivity or interference between Rat CX31 and analogues was observed.
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 0.63 to 40 ng/mL
Assay Sensitivity 0.38 ng/mL
Product Type ELISA Kit (Colorimetric)
Sample Type Other Biological Fluids, Plasma, Serum
For Use With (Application) ELISA
Gene ID (Entrez) 2707
Gene Symbol GJB3
Host Species Rat
Quantity 1 Kit
Storage Requirements Storage of components varies. See protocol for specific instructions.
Target Connexin 31/GJB3
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.