missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLC4A1AP Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA599237
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total NADAP.
SLC4A1AP gene ontology (GO) annotation includes protein binding and plasma membrane.
Specifications
| SLC4A1AP | |
| Polyclonal | |
| Unconjugated | |
| SLC4A1AP | |
| HLC3; HLC-3; Human lung cancer oncogene 3 protein; kanadaptin; Kidney anion exchanger adapter protein; kidney anion exchanger adaptor protein; lung cancer oncogene 3; SLC4A1AP; solute carrier family 4 (anion exchanger), member 1, adapter protein; solute carrier family 4 (anion exchanger), member 1, adaptor protein; solute carrier family 4 anion exchanger member 1 adapter protein; solute carrier family 4 member 1 adaptor protein | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 20534, 22950 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9BWU0 | |
| SLC4A1AP | |
| A synthesized peptide derived from human NADAP, corresponding to a region within C-terminal amino acids. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction