missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLC52A3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5113391
This item is not returnable.
View return policy
Description
SLC52A3 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA
This gene encodes a riboflavin transporter protein that is strongly expressed in the intestine and likely plays a role in intestinal absorption of riboflavin. The protein is predicted to have eleven transmembrane domains and a cell surface localization signal in the C-terminus. Mutations at this locus have been associated with Brown-Vialetto-Van Laere syndrome and Fazio-Londe disease. [provided by RefSeq, Mar 2012].
Specifications
| SLC52A3 | |
| Polyclonal | |
| Unconjugated | |
| SLC52A3 | |
| 2310046K01Rik; bA371L19.1; BVVLS; BVVLS1; C20orf54; ct054; hRFT2; Rft2; RFVT3; RGD1304644; Riboflavin transporter 2; rRFT2; Slc52a3; solute carrier family 52 (riboflavin transporter), member 3; solute carrier family 52 member 3; solute carrier family 52, riboflavin transporter, member 3; solute carrier protein family 52, member 3 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 113278, 69698 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q9D6X5, Q9NQ40 | |
| SLC52A3 | |
| Recombinant Human Solute carrier family 52, riboflavin transporter, member 3 protein (159-220AA). | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction