Learn More
Invitrogen™ SLC5A3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA533041
Description
Percent identity with other species by BLAST analysis: Human, Baboon, Monkey, Galago, Bat, Bovine, Dog, Elephant, Panda, Rabbit, Pig (100%); Marmoset, Mouse, Hamster, Armadillo (94%); Shrew (89%); Rat (83%). For IHC(P), use heat induced antigen retrieval in pH 6.0 citrate buffer. After incubation with the primary antibody, slides were incubated with biotinylated secondary antibody, followed by alkaline phosphatase-streptavidin and chromogen.
Prevents intracellular accumulation of high concentrations of myo-inositol (an osmolyte) that result in impairment of cellular function.
Specifications
| SLC5A3 | |
| Polyclonal | |
| Unconjugated | |
| SLC5A3 | |
| AA623876; BCW2; BCW2 protein; BF642829; Na(+)/myo-inositol cotransporter; Slc5a3; SMIT; Smit1; SMIT2; sodium myo-inositol transporter 1; sodium/myo-inositol cotransporter; sodium/myo-inositol cotransporter 1; sodium/myo-inositol transporter 1; solute carrier family 5 (inositol transporters), member 3; solute carrier family 5 (sodium/myo-inositol cotransporter), member 3; solute carrier family 5 member 3; solute carrier family 5, member 3; solute carrier family 5, member alpha 3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 100328816, 106558089, 110255177, 282362, 445542, 6526 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| P31637, P53793, P53794 | |
| SLC5A3 | |
| Synthetic 18 amino acid peptide from C-terminus of human SLC5A3. | |
| 50 μg | |
| Primary | |
| Human, Canine, Rabbit, Bovine, Pig, Bat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.