Learn More
Invitrogen™ SLC5A5 Monoclonal Antibody (14F), FITC
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  Invitrogen™ MA545490
Product Code. 17845822
  • € 467.00 / 100µg
Description
1 μg/mL of MA5-45490 was sufficient for detection of hNIS in 20 μg of transfected COS-7 cell membrane lysate by ECL immunoblot analysis using Goat anti-mouse IgG:HRP as the secondary antibody. Detects approximately 97kDa, non-glycosylated version at 68kDa. Other minor bands associated with hNIS at 160kDa, and degradation products at approximately 30 kDa, and approximately 15kDa.
Human Sodium Iodide Symporter (hNIS) is responsible for iodide concentrating ability within thyroid follicular cells. It is a membrane bound glycoprotein with 13 membrane spanning domains and 14 extramembranous domains. It may represent an autoantigen in thyroid.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SLC5A5
Monoclonal
1 mg/mL
PBS, 9.1mM sodium bicarbonate with 640.91mM DMSO, 136.36mM ethanolamine and no preservative; pH 7.4
Q63008, Q92911, Q99PN0
SLC5A5
Mannose binding protein hNIS fusion (AA468-643).
100 μg
Primary
Human, Mouse, Rat
Antibody
IgG1
Immunohistochemistry (PFA fixed), Western Blot, Microarray
14F
FITC
SLC5A5
Na(+)/I(-) cotransporter; Na(+)/I(-) symporter; Na(+)/I(-)-symporter; Na+/I- symporter; Nis; SLC5A5; sodium iodide symporter; Sodium/iodide cotransporter; sodium-iodide symporter; solute carrier family 5 (sodium iodide symporter), member 5; solute carrier family 5 (sodium/iodide cotransporter), member 5; solute carrier family 5 member 5; solute carrier family 5, member 5; TDH1
Mouse
Protein G
RUO
114479, 114613, 6528
4°C, store in dark