missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC5A5/Sodium Iodide Symporter Antibody (14F), Janelia Fluor™ 646, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59341JF646
This item is not returnable.
View return policy
Description
SLC5A5/Sodium Iodide Symporter Monoclonal antibody specifically detects SLC5A5/Sodium Iodide Symporter in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Microarray
Specifications
| SLC5A5/Sodium Iodide Symporter | |
| Monoclonal | |
| Janelia Fluor 646 | |
| Na(+)/I(-) cotransporter, Na(+)/I(-) symporter, NISNa(+)/I(-)-symporter, sodium/iodide cotransporter, Sodium-iodide symporter, solute carrier family 5 (sodium iodide symporter), member 5, Solute carrier family 5 member 5, TDH1 | |
| Mannose binding protein hNIS fusion (AA468-643) | |
| 0.1 mL | |
| ABC Transporters, Cancer, Cellular Signaling, Neuroscience | |
| 6528 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Immunohistochemistry, Microarray | |
| 14F | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction