missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
SLC5A5/Sodium Iodide Symporter Monoclonal specifically detects SLC5A5/Sodium Iodide Symporter in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
| Antigen | SLC5A5/Sodium Iodide Symporter |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | FP5 |
| Conjugate | HRP |
| Dilution | Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin |
| Formulation | PBS with No Preservative |
| Gene Accession No. | W9JM47 |
| Gene Alias | Na(+)/I(-) cotransporter, Na(+)/I(-) symporter, NISNa(+)/I(-)-symporter, sodium/iodide cotransporter, Sodium-iodide symporter, solute carrier family 5 (sodium iodide symporter), member 5, Solute carrier family 5 member 5, TDH1 |
| Gene Symbols | SLC5A5 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?