missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLC5A8 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5113782
This item is not returnable.
View return policy
Description
Immunogen sequence: CNSTYNETNL MTTTEMPFTT SVFQIYNVQR TPLMDNWYSL SYLYFSTVGT For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Human: 100%.
SLC5A8 has been shown to transport iodide by a passive mechanism (Rodriguez et al., 2002 ) and monocarboxylates and short-chain fatty acids by a sodium-coupled mechanism (Gopal et al., 2004 ).
Specifications
| SLC5A8 | |
| Polyclonal | |
| Unconjugated | |
| Slc5a8 | |
| AIT; Apical iodide transporter; apical iodide transporter , putative; electrogenic sodium monocarboxylate cotransporter; RGD1564146; Slc5a8; SMCT; SMCT1; Sodium iodide-related cotransporter; sodium-coupled monocarboxylate transporter 1; solute carrier family 5 (iodide transporter), member 8; solute carrier family 5 (sodium/monocarboxylate cotransporter), member 8; solute carrier family 5 member 8 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 160728 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8N695 | |
| Slc5a8 | |
| Synthetic peptide directed towards the C-terminal region of Human SLC5A8 (aa 480-529). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction