missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLC5A9 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA534212
This item is not returnable.
View return policy
Description
Percent identity with other species by BLAST analysis: Human, Chimpanzee, Gorilla, Orangutan, Gibbon, Monkey, Marmoset (100%); Elephant, Dog (94%); Galago, Panda, Bovine, Horse, Pig (88%); Mouse, Bat (82%). Proteinase K treatment is required for antigen retrieval.
Involved in sodium-dependent transport of D-mannose, D-glucose and D-fructose.
Specifications
| SLC5A9 | |
| Polyclonal | |
| Unconjugated | |
| SLC5A9 | |
| hSGLT4; Na(+)/glucose cotransporter 4; SGLT4; SLC5A9; Sodium/glucose cotransporter 4; solute carrier family 5 (sodium/glucose cotransporter), member 9; solute carrier family 5 (sodium/sugar cotransporter), member 9; solute carrier family 5 member 9 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 200010 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| Q2M3M2 | |
| SLC5A9 | |
| Synthetic 17 amino acid peptide from internal region of human SLC5A9. | |
| 50 μg | |
| Primary | |
| Human, Monkey | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction