missing translation for 'onlineSavingsMsg'
Learn More

SLC6A18 Rabbit Polyclonal Antibody, Proteintech

Rabbit Polyclonal Antibody

€ 145.00 - € 410.00

Specifications

Antigen SLC6A18
Concentration 300 μg/mL
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30237507
View Documents
Proteintech
17876-1-AP-20UL
20 μL
€ 145.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30237993
View Documents
Proteintech
17876-1-AP-150UL
150 μL
€ 410.00
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The SLC6A18 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human SLC6A18. This antibody recognizes human, mouse antigen. The SLC6A18 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications

Specifications

SLC6A18
Western Blot, ELISA
Unconjugated
Rabbit
Human, Mouse
FLJ31236; Xtrp2
IgG
Antigen affinity purification
Polyclonal Antibody
300 μg/mL
Polyclonal
Liquid
RUO
348932
Recombinant Protein
Primary
Store at -20°C. Stable for one year after shipment.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.