Learn More
SLC6A3/DAT1 Antibody (CL3123) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-43803
Product Code. 30287617
  • € 561.00 / 100µL
Description
SLC6A3/DAT1 Monoclonal antibody specifically detects SLC6A3/DAT1 in Human,Mouse,Rat samples. It is validated for Immunohistochemistry (Paraffin),Immunohistochemistry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SLC6A3/DAT1
Monoclonal
LYOPH
Immunohistochemistry-Paraffin 1:200 - 1:500
DA transporter, DATDAT1Solute carrier family 6 member 3, sodium-dependent dopamine transporter, solute carrier family 6 (neurotransmitter transporter, dopamine), member 3
This antibody was generated using a recombinant protein sequence of Q01959, with the exact immunogen sequence remaining proprietary.
100 μL
Neuroscience
6531
Human, Mouse, Rat
Purified
Immunohistochemistry (Paraffin), Immunohistochemistry
CL3123
Unconjugated
Lyophilized in 5% Trehalose.
Mouse
Protein A purified
RUO
Primary
Centrifuge the vial of lyophilized antibody at 12,000 x g for 20 seconds. Reconstitute by adding sterile, distilled water to achieve a final antibody concentration of 1mg/mL.
Store at 4⪚C short term. Aliquot and store at -20⪚C long term. Avoid freeze-thaw cycles.