missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC6A3/DAT1 Antibody (CL3123) [Biotin], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-46649B
This item is not returnable.
View return policy
Description
SLC6A3/DAT1 Monoclonal antibody specifically detects SLC6A3/DAT1 in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
Specifications
| SLC6A3/DAT1 | |
| Monoclonal | |
| Biotin | |
| DA transporter, DATDAT1Solute carrier family 6 member 3, sodium-dependent dopamine transporter, solute carrier family 6 (neurotransmitter transporter, dopamine), member 3 | |
| This antibody was developed using a recombinant protein derived from Q01959, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Neuroscience | |
| 6531 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| CL3123 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction