missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC6A4/5-HTTLPR/Serotonin transporter Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NB100-1892
This item is not returnable.
View return policy
Description
SLC6A4/5-HTTLPR/Serotonin transporter Polyclonal specifically detects SLC6A4/5-HTTLPR/Serotonin transporter in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Peptide ELISA.
Specifications
| SLC6A4/5-HTTLPR/Serotonin transporter | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| 5HT transporter, 5-HTT, hSERT, HTT5-hydroxytryptamine transporter, Na+/Cl- dependent serotonin transporter, OCD1, SERT, sodium-dependent serotonin transporter, solute carrier family 6 (neurotransmitter transporter, serotonin), member 4,5-HTTLPR, Solute carrier family 6 member 4,5HTT | |
| Goat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Expected from sequence similarity: Rat, Mouse. | |
| Store at -20C. Avoid freeze-thaw cycles. |
| Western Blot, Immunocytochemistry, Immunofluorescence, ELISA | |
| 0.5 mg/mL | |
| Western Blot 1 - 3 ug/mL, Immunocytochemistry/Immunofluorescence 10 ug/mL, Peptide ELISA Detection limit 1:32000 | |
| NP_001036.1. | |
| SLC6A4 | |
| Peptide with sequence C-LHQGERETWGK corresponding to internal region according to NP_001036.1. | |
| 0.1 mg | |
| Neuroscience | |
| 6532 | |
| Human, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction